Assay Details
Target Gene Details
Entrez Gene ID: | 4649 |
Gene Name: | myosin IXA |
Gene Aliases: |
- |
Location: |
Chr.15:71822290-72118577 on Build GRCh38 |
Assay Gene Location: | Within Intron 45 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892712 | Chr.15:71809756 - 72033254 on Build GRCh38 | Loss |
|
| nsv1036938 | Chr.15:71655767 - 71980514 on Build GRCh38 | Gain |
|
| esv2531029 | Chr.15:71831939 - 71833474 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map