Assay Details
Target Gene Details
Entrez Gene ID: | 116179 |
Gene Name: | transglutaminase 7 |
Gene Aliases: |
TGMZ |
Location: |
Chr.15:43276277-43302255 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGM7 |

Genomic Map