Assay Details
Target Gene Details
Entrez Gene ID: | 11261 |
Gene Name: | calcineurin like EF-hand protein 1 |
Gene Aliases: |
CHP, SLC9A1BP, Sid470p, p22, p24 |
Location: |
Chr.15:41231149-41281887 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHP1 | NM_007236.4 | NP_009167.1 | ||
| XM_017021879.1 | XP_016877368.1 | |||
| AK094363.1 | 1 | 816 | ||
| AK299660.1 | ||||
| AK307060.1 | ||||
| AK312582.1 | ||||
| BC031293.1 | AAH31293.1 | |||
| CR536539.1 | CAG38776.1 | |||
| CR542085.1 | CAG46882.1 | |||
| U61538.1 | AAB37770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045455 | Chr.15:41244426 - 41328336 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map