Assay Details
Target Gene Details
Entrez Gene ID: | 5597 |
Gene Name: | mitogen-activated protein kinase 6 |
Gene Aliases: |
ERK3, HsT17250, PRKM6, p97MAPK |
Location: |
Chr.15:52004265-52066265 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK6 | NM_002748.3 | NP_002739.1 | ||
| XM_005254537.2 | XP_005254594.1 | |||
| XM_005254538.2 | XP_005254595.1 | |||
| XM_005254539.3 | XP_005254596.1 | |||
| XM_011521782.1 | XP_011520084.1 | |||
| BC035492.2 | AAH35492.1 | |||
| CR749401.1 | CAH18246.1 | |||
| DA109756.1 | ||||
| L77964.1 | AAA98769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2595n100 | Chr.15:51964903 - 52049904 on Build GRCh38 | Gain |
|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map