Assay Details
Target Gene Details
Entrez Gene ID: | 10518 |
Gene Name: | calcium and integrin binding family member 2 |
Gene Aliases: |
DFNB48, KIP2, USH1J |
Location: |
Chr.15:78104606-78131976 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB2 | NM_001271888.1 | NP_001258817.1 | ||
| NM_001271889.1 | NP_001258818.1 | |||
| NM_001301224.1 | NP_001288153.1 | |||
| NM_006383.3 | NP_006374.1 | |||
| NR_125435.1 | ||||
| XM_005254126.3 | XP_005254183.1 | |||
| XM_006720374.2 | XP_006720437.1 | |||
| XM_011521161.2 | XP_011519463.1 | |||
| AB012955.1 | BAA33584.1 | |||
| AI817087.1 | ||||
| AI857912.1 | ||||
| AK293167.1 | ||||
| BC033108.1 | ||||
| BC047381.1 | AAH47381.1 | |||
| BE042882.1 | ||||
| BT019962.1 | AAV38765.1 | |||
| BU556614.1 | ||||
| CN371150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581778 | Chr.15:78106304 - 78117814 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map