Assay Details
Target Gene Details
Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049980 | Chr.15:67241206 - 67285222 on Build GRCh38 | Loss |
|

Genomic Map