Assay Details
Target Gene Details
Entrez Gene ID: | 3712 |
Gene Name: | isovaleryl-CoA dehydrogenase |
Gene Aliases: |
ACAD2 |
Location: |
Chr.15:40405485-40435948 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IVD | XM_005254350.2 | XP_005254407.1 | ||
| XM_006720492.2 | XP_006720555.1 | |||
| XM_006720495.2 | XP_006720558.1 | |||
| XM_017022149.1 | XP_016877638.1 | |||
| XM_017022150.1 | XP_016877639.1 | |||
| XM_017022151.1 | XP_016877640.1 | |||
| XM_017022153.1 | XP_016877642.1 | |||
| XM_017022157.1 | XP_016877646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510660 | Chr.15:40260705 - 40429684 on Build GRCh38 | Deletion |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map