Assay Details
Target Gene Details
Entrez Gene ID: | 54962 |
Gene Name: | TIMELESS interacting protein |
Gene Aliases: |
- |
Location: |
Chr.15:66334976-66386787 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIPIN | XM_005254521.2 | XP_005254578.1 | ||
| XM_017022390.1 | XP_016877879.1 | |||
| XM_017022391.1 | XP_016877880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047588 | Chr.15:66227169 - 66394432 on Build GRCh38 | Loss |
|
| nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map