Assay Details
Target Gene Details
Entrez Gene ID: | 105370792 |
Gene Name: | uncharacterized LOC105370792 |
Gene Aliases: |
- |
Location: |
Chr.15:41892777-41898815 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105370792 | NR_135681.1 | 3 | 624 | |
| AK057625.1 | 2 | 2756 | ||
| BC022404.1 | 1 | 807 | AAH22404.1 | |
| DA112916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain |
|
| nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain |
|
| dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain |
|

Genomic Map