Assay Details
Target Gene Details
Entrez Gene ID: | 3681 |
Gene Name: | integrin subunit alpha D |
Gene Aliases: |
ADB2, CD11D |
Location: |
Chr.16:31393224-31426513 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGAD | NM_001318185.1 | NP_001305114.1 | ||
| NM_005353.2 | NP_005344.2 | |||
| XM_011545835.2 | XP_011544137.1 | |||
| XM_011545836.2 | XP_011544138.1 | |||
| XM_011545837.2 | XP_011544139.1 | |||
| XM_011545838.2 | XP_011544140.1 | |||
| XM_011545839.2 | XP_011544141.1 | |||
| XM_011545841.2 | XP_011544143.1 | |||
| XM_011545842.2 | XP_011544144.1 | |||
| XM_011545843.2 | XP_011544145.1 | |||
| XM_011545844.2 | XP_011544146.1 | |||
| XM_011545845.2 | XP_011544147.1 | |||
| XM_011545846.2 | XP_011544148.1 | |||
| XM_011545847.2 | XP_011544149.1 | |||
| XM_011545848.2 | XP_011544150.1 | |||
| XM_017023215.1 | XP_016878704.1 | |||
| AB208945.1 | BAD92182.1 | |||
| AK097160.1 | BAC04967.1 | |||
| AK301028.1 | ||||
| U37028.1 | AAB38547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714194 | Chr.16:31362156 - 31410550 on Build GRCh38 | Deletion |
|
| esv3638400 | Chr.16:31404264 - 31407669 on Build GRCh38 | Loss |
|
| nsv1059610 | Chr.16:31357378 - 31410339 on Build GRCh38 | Gain |
|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| nsv1066280 | Chr.16:31385143 - 31440746 on Build GRCh38 | Gain |
|
| dgv503e214 | Chr.16:31332171 - 31413104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map