Assay Details
Target Gene Details
Entrez Gene ID: | 54496 |
Gene Name: | protein arginine methyltransferase 7 |
Gene Aliases: |
- |
Location: |
Chr.16:68310974-68358832 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
More Information
Set Membership: |
|

Genomic Map