Assay Details
Target Gene Details
Entrez Gene ID: | 3683 |
Gene Name: | integrin subunit alpha L |
Gene Aliases: |
CD11A, LFA-1, LFA1A |
Location: |
Chr.16:30472662-30523185 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGAL | NM_001114380.1 | NP_001107852.1 | ||
| NM_002209.2 | NP_002200.2 | |||
| XM_005255313.1 | XP_005255370.1 | |||
| XM_006721044.1 | XP_006721107.1 | |||
| XM_011545849.1 | XP_011544151.1 | |||
| AK298674.1 | ||||
| AK303190.1 | ||||
| AK314246.1 | ||||
| BC008777.2 | AAH08777.1 | |||
| Y00796.1 | CAA68747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1783 | Chr.16:30438129 - 30483000 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map