Assay Details
Target Gene Details
Entrez Gene ID: | 51741 |
Gene Name: | WW domain containing oxidoreductase |
Gene Aliases: |
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1 |
Location: |
Chr.16:78099413-79212667 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWOX | NM_001291997.1 | NP_001278926.1 | ||
| NM_016373.3 | NP_057457.1 | |||
| NM_130791.3 | NP_570607.1 | |||
| NR_120436.1 | ||||
| XM_011523101.2 | XP_011521403.1 | |||
| XM_011523103.2 | XP_011521405.1 | |||
| XM_011523104.2 | XP_011521406.1 | |||
| XM_011523105.2 | XP_011521407.1 | |||
| XM_017023278.1 | XP_016878767.1 | |||
| AF187015.1 | AAF31694.1 | |||
| AF211943.1 | AAF27049.1 | |||
| AF227526.1 | AAF82053.1 | |||
| AF227527.1 | AAF82054.1 | |||
| AF227528.1 | AAF82055.1 | |||
| AF395123.1 | AAK81727.1 | |||
| AF395124.1 | AAK81728.1 | |||
| AK027626.1 | ||||
| AK290438.1 | ||||
| AK291300.1 | ||||
| AK298322.1 | ||||
| AW874692.1 | ||||
| AY256821.1 | AAP94227.1 | |||
| BC003184.1 | AAH03184.1 | |||
| BU157392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv539e214 | Chr.16:78117264 - 78142432 on Build GRCh38 | Loss |
|
| dgv3031n100 | Chr.16:77892138 - 78561070 on Build GRCh38 | Gain |
|
| nsv1056016 | Chr.16:77990656 - 78129245 on Build GRCh38 | Loss |
|
| nsv833293 | Chr.16:77932757 - 78134791 on Build GRCh38 | Gain |
|
| esv2760418 | Chr.16:77894064 - 78560274 on Build GRCh38 | Gain+Loss |
|
| esv3892902 | Chr.16:78049580 - 78674801 on Build GRCh38 | Gain |
|
| nsv573133 | Chr.16:78078366 - 78181055 on Build GRCh38 | Gain |
|
| nsv520408 | Chr.16:78117849 - 78137411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map