Assay Details
Target Gene Details
Entrez Gene ID: | 79650 |
Gene Name: | U6 snRNA biogenesis phosphodiesterase 1 |
Gene Aliases: |
C16orf57, HVSL1, Mpn1, PN, hUsb1 |
Location: |
Chr.16:57999600-58021623 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USB1 | XM_005256144.3 | XP_005256201.1 | ||
| XM_011523329.2 | XP_011521631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv29242 | Chr.16:58000150 - 58000829 on Build GRCh38 | Loss |
|
| nsv1063386 | Chr.16:57958821 - 58040306 on Build GRCh38 | Gain |
|
| nsv1816 | Chr.16:57973452 - 58018171 on Build GRCh38 | Deletion |
|
| nsv572759 | Chr.16:57972385 - 58040306 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map