Assay Details
Target Gene Details
Entrez Gene ID: | 23059 |
Gene Name: | clusterin associated protein 1 |
Gene Aliases: |
CFAP22, FAP22, IFT38 |
Location: |
Chr.16:3495562-3539048 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLUAP1 | NM_015041.2 | 12 | 3952 | NP_055856.1 |
| NM_024793.2 | 8 | 3410 | NP_079069.1 | |
| BQ446386.1 | 1 | 207 |
Target Gene Details
Entrez Gene ID: | 197358 |
Gene Name: | NLR family CARD domain containing 3 |
Gene Aliases: |
CLR16.2, NOD3 |
Location: |
Chr.16:3538272-3578373 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRC3 | XM_017023027.1 | 20 | 6619 | XP_016878516.1 |
| XM_017023028.1 | 21 | 6793 | XP_016878517.1 | |
| XM_017023029.1 | 22 | 6874 | XP_016878518.1 | |
| XM_017023030.1 | 22 | 6935 | XP_016878519.1 | |
| XM_017023031.1 | 20 | 6680 | XP_016878520.1 | |
| XM_017023032.1 | 22 | 7636 | XP_016878521.1 | |
| XM_017023033.1 | 22 | 6813 | XP_016878522.1 | |
| XM_017023034.1 | 21 | 6732 | XP_016878523.1 | |
| XM_017023035.1 | 21 | 6761 | XP_016878524.1 | |
| XM_017023036.1 | 20 | 7320 | XP_016878525.1 | |
| XM_017023037.1 | 21 | 6514 | XP_016878526.1 | |
| XM_017023038.1 | 21 | 6487 | XP_016878527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| nsv1716 | Chr.16:3511573 - 3544041 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map