Assay Details
Target Gene Details
Entrez Gene ID: | 6361 |
Gene Name: | C-C motif chemokine ligand 17 |
Gene Aliases: |
A-152E5.3, ABCD-2, SCYA17, TARC |
Location: |
Chr.16:57396076-57416063 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL17 | NM_002987.2 | NP_002978.1 | ||
| XM_011523256.2 | XP_011521558.1 | |||
| XM_017023530.1 | XP_016879019.1 | |||
| BC069107.1 | AAH69107.1 | |||
| BC112066.1 | AAI12067.1 | |||
| BC112068.1 | AAI12069.1 | |||
| D43767.1 | BAA07824.1 |
More Information
Set Membership: |
|

Genomic Map