Assay Details
Target Gene Details
Entrez Gene ID: | 197370 |
Gene Name: | NSE1 homolog, SMC5-SMC6 complex component |
Gene Aliases: |
NSE1 |
Location: |
Chr.16:27224994-27268810 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSMCE1 | NM_145080.3 | NP_659547.2 | ||
| XM_006721023.3 | XP_006721086.1 | |||
| XM_011545761.2 | XP_011544063.1 | |||
| XM_011545762.1 | XP_011544064.1 | |||
| XM_017023040.1 | XP_016878529.1 | |||
| XM_017023041.1 | XP_016878530.1 | |||
| XM_017023042.1 | XP_016878531.1 | |||
| AF161451.1 | AAF29011.1 | |||
| AF161455.1 | AAF29015.1 | |||
| AK056795.1 | ||||
| AK315674.1 | ||||
| BC018938.2 | ||||
| DB463393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059454 | Chr.16:27139937 - 27679877 on Build GRCh38 | Gain |
|
| esv3638319 | Chr.16:27201260 - 27313624 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map