Assay Details
Target Gene Details
Entrez Gene ID: | 5336 |
Gene Name: | phospholipase C gamma 2 |
Gene Aliases: |
APLAID, FCAS3, PLC-IV, PLC-gamma-2 |
Location: |
Chr.16:81779258-81962693 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCG2 | NM_002661.4 | NP_002652.2 | ||
| AB208914.1 | BAD92151.1 | |||
| AK310042.1 | ||||
| AY364256.1 | AAQ76815.1 | |||
| BC007565.1 | AAH07565.1 | |||
| BC011772.2 | AAH11772.1 | |||
| BC014561.2 | AAH14561.1 | |||
| BC018646.2 | AAH18646.1 | |||
| BP351485.1 | ||||
| BX504887.1 | ||||
| DB456860.1 | ||||
| M37238.1 | AAA60112.1 | |||
| X14034.1 | CAA32194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639343 | Chr.16:81755626 - 81815373 on Build GRCh38 | Loss |
|
| nsv573365 | Chr.16:81771367 - 81942287 on Build GRCh38 | Loss |
|
| esv3639339 | Chr.16:81696019 - 81869162 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80343087] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map