Assay Details
Target Gene Details
Entrez Gene ID: | 146198 |
Gene Name: | ZFP90 zinc finger protein |
Gene Aliases: |
FIK, NK10, ZNF756, zfp-90 |
Location: |
Chr.16:68533484-68576072 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP90 | XM_017022952.1 | XP_016878441.1 |
More Information
Set Membership: |
|

Genomic Map