Assay Details
Target Gene Details
Entrez Gene ID: | 23059 |
Gene Name: | clusterin associated protein 1 |
Gene Aliases: |
CFAP22, FAP22, IFT38 |
Location: |
Chr.16:3495562-3539048 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLUAP1 | NM_015041.2 | NP_055856.1 | ||
| NM_024793.2 | NP_079069.1 | |||
| XM_006720867.3 | XP_006720930.1 | |||
| XM_017023068.1 | XP_016878557.1 | |||
| XM_017023069.1 | XP_016878558.1 | |||
| XM_017023070.1 | XP_016878559.1 | |||
| XM_017023071.1 | XP_016878560.1 | |||
| XM_017023072.1 | XP_016878561.1 | |||
| XM_017023073.1 | XP_016878562.1 | |||
| AB014543.1 | BAA31618.1 | |||
| AB089691.1 | BAD44779.1 | |||
| AK023319.1 | BAB14523.1 | |||
| AK023359.1 | BAB14542.1 | |||
| AK294318.1 | ||||
| BC017070.2 | AAH17070.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| nsv1716 | Chr.16:3511573 - 3544041 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140121457] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map