Assay Details
Target Gene Details
Entrez Gene ID: | 865 |
Gene Name: | core-binding factor, beta subunit |
Gene Aliases: |
PEBP2B |
Location: |
Chr.16:67029147-67101058 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBFB | NM_001755.2 | NP_001746.1 | ||
| NM_022845.2 | NP_074036.1 | |||
| XM_005256212.3 | XP_005256269.1 | |||
| XM_006721321.2 | XP_006721384.1 | |||
| XM_017023812.1 | XP_016879301.1 | |||
| XM_017023813.1 | XP_016879302.1 | |||
| AA020956.1 | ||||
| AF294326.1 | AAG01553.1 | |||
| AK290462.1 | ||||
| AK291834.1 | ||||
| AK311724.1 | ||||
| BC018509.1 | AAH18509.1 | |||
| BM679848.1 | ||||
| BT006862.1 | AAP35508.1 | |||
| L20298.1 | AAA02868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516064 | Chr.16:67039649 - 67091161 on Build GRCh38 | Gain |
|
| nsv1056738 | Chr.16:67021320 - 67091992 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map