Assay Details
Target Gene Details
Entrez Gene ID: | 51573 |
Gene Name: | glycerophosphodiester phosphodiesterase 1 |
Gene Aliases: |
363E6.2, MIR16 |
Location: |
Chr.16:19501693-19522145 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDE1 | NM_001324066.1 | NP_001310995.1 | ||
| NM_001324067.1 | NP_001310996.1 | |||
| NM_016641.3 | NP_057725.1 | |||
| NR_136689.1 | ||||
| AF212862.1 | AAF65234.1 | |||
| AF306698.1 | ||||
| AK312912.1 | ||||
| AY463154.1 | AAR25624.1 | |||
| BC014981.1 | AAH14981.1 | |||
| BC025273.1 | AAH25273.1 | |||
| BQ897064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833163 | Chr.16:19457512 - 19628716 on Build GRCh38 | Gain |
|
| nsv571582 | Chr.16:19038056 - 19790801 on Build GRCh38 | Gain |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map