Assay Details
Target Gene Details
Entrez Gene ID: | 2775 |
Gene Name: | G protein subunit alpha o1 |
Gene Aliases: |
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1 |
Location: |
Chr.16:56191339-56357444 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAO1 | NM_020988.2 | NP_066268.1 | ||
| NM_138736.2 | NP_620073.2 | |||
| XM_011523003.2 | XP_011521305.1 | |||
| AF493894.1 | AAM12608.1 | |||
| AF493895.1 | AAM12609.1 | |||
| AK056008.1 | ||||
| BC030027.2 | AAH30027.2 | |||
| CR936770.1 | ||||
| KR081949.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117267531] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map