Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSE1 | XM_005255859.4 | XP_005255916.3 | ||
| XM_005255860.3 | XP_005255917.3 | |||
| XM_005255861.4 | XP_005255918.3 |
Target Gene Details
Entrez Gene ID: | 197196 |
Gene Name: | long intergenic non-protein coding RNA 311 |
Gene Aliases: |
NCRNA00311, TMEM148 |
Location: |
Chr.16:85282958-85288082 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00311 | NR_038859.1 | |||
| BC030801.1 | AAH30801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063498 | Chr.16:85218793 - 85287509 on Build GRCh38 | Loss |
|
| nsv833313 | Chr.16:85282402 - 85458317 on Build GRCh38 | Loss |
|
| nsv952062 | Chr.16:85245895 - 85668994 on Build GRCh38 | Deletion |
|
| esv3639461 | Chr.16:85277814 - 85284408 on Build GRCh38 | Loss |
|
| nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss |
|
| esv2677829 | Chr.16:85281196 - 85284462 on Build GRCh38 | Deletion |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map