Assay Details
Target Gene Details
Entrez Gene ID: | 29117 |
Gene Name: | bromodomain containing 7 |
Gene Aliases: |
BP75, CELTIX1, NAG4 |
Location: |
Chr.16:50317563-50368969 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRD7 | NM_001173984.2 | NP_001167455.1 | ||
| NM_013263.4 | NP_037395.2 | |||
| XM_011523046.2 | XP_011521348.1 | |||
| XM_011523047.2 | XP_011521349.1 | |||
| XM_011523048.2 | XP_011521350.1 | |||
| XM_011523049.2 | XP_011521351.1 | |||
| XM_011523050.2 | XP_011521352.1 | |||
| XM_017023179.1 | XP_016878668.1 | |||
| XM_017023180.1 | XP_016878669.1 | |||
| XM_017023181.1 | XP_016878670.1 | |||
| AF152604.1 | AAF75126.1 | |||
| AF213969.1 | AAF19526.1 | |||
| AJ271881.1 | CAB72445.1 | |||
| AK027308.1 | BAB55031.1 | |||
| AK074613.1 | BAC11089.1 | |||
| BC001611.1 | AAH01611.1 | |||
| BC050728.1 | AAH50728.1 | |||
| BC094706.1 | AAH94706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv978324 | Chr.16:50325790 - 50328674 on Build GRCh38 | Duplication |
|
| nsv1805 | Chr.16:50307115 - 50352023 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140047531] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map