Assay Details
Target Gene Details
Entrez Gene ID: | 164 |
Gene Name: | adaptor related protein complex 1 gamma 1 subunit |
Gene Aliases: |
ADTG, CLAPG1 |
Location: |
Chr.16:71729000-71809073 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1G1 | NM_001030007.1 | NP_001025178.1 | ||
| NM_001128.5 | NP_001119.3 | |||
| AB015317.1 | BAA33389.1 | |||
| AK055122.1 | ||||
| AK128078.1 | ||||
| AK225906.1 | ||||
| AK293233.1 | ||||
| AK314484.1 | ||||
| AL110198.1 | CAB53673.1 | |||
| BC036283.1 | AAH36283.1 | |||
| Y12226.1 | CAA72902.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76479594] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map