Assay Details
Target Gene Details
Entrez Gene ID: | 10294 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A2 |
Gene Aliases: |
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2 |
Location: |
Chr.16:46955362-46973714 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA2 | NM_005880.3 | NP_005871.1 | ||
| AJ001309.1 | CAA04669.1 | |||
| AK313031.1 | ||||
| BC013044.2 | AAH13044.1 | |||
| BC015809.2 | AAH15809.1 | |||
| Y13350.1 | CAA73791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map