Assay Details
Target Gene Details
Entrez Gene ID: | 81533 |
Gene Name: | integrin alpha FG-GAP repeat containing 1 |
Gene Aliases: |
2310047C21Rik, CDA08, LNKN-1, TIP |
Location: |
Chr.16:47154387-47461274 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITFG1 | NM_001305002.1 | NP_001291931.1 | ||
| NM_030790.4 | NP_110417.2 | |||
| AF212247.1 | AAG41782.1 | |||
| AF503339.1 | AAN31655.1 | |||
| AK027596.1 | BAB55220.1 | |||
| AK295228.1 | ||||
| AK299641.1 | ||||
| AK301908.1 | ||||
| AL833705.1 | ||||
| BC006321.2 | AAH06321.2 | |||
| BC024162.1 | AAH24162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
| nsv528423 | Chr.16:47129963 - 47313869 on Build GRCh38 | Loss |
|
| nsv525170 | Chr.16:47160140 - 47269724 on Build GRCh38 | Gain |
|

Genomic Map