Assay Details
Target Gene Details
Entrez Gene ID: | 115 |
Gene Name: | adenylate cyclase 9 |
Gene Aliases: |
AC9, ACIX |
Location: |
Chr.16:3953387-4116185 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY9 | XM_011522353.2 | XP_011520655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21490 | Chr.16:3825338 - 4270692 on Build GRCh38 | Gain+Loss |
|
| nsv1038196 | Chr.16:3917410 - 4153984 on Build GRCh38 | Gain |
|
| nsv833130 | Chr.16:3810372 - 3975509 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76884330] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map