Assay Details
Target Gene Details
Entrez Gene ID: | 10204 |
Gene Name: | nuclear transport factor 2 |
Gene Aliases: |
NTF-2, NTF2, PP15 |
Location: |
Chr.16:67846732-67872567 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUTF2 | NM_001322038.1 | 6 | 1879 | NP_001308967.1 |
| NM_001322039.1 | 6 | 1968 | NP_001308968.1 | |
| NM_001322040.1 | 5 | 1878 | NP_001308969.1 | |
| NM_001322041.1 | 5 | 1988 | NP_001308970.1 | |
| NM_005796.2 | 5 | 1789 | NP_005787.1 | |
| BX647782.1 | 1 | 2076 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv1844 | Chr.16:67868325 - 67917356 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map