Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYM | NM_001888.4 | NP_001879.1 | ||
| AK290852.1 | ||||
| AK310424.1 | ||||
| BC018061.1 | AAH18061.1 | |||
| BX648477.1 | CAI46030.1 | |||
| L02950.1 | AAC16914.1 | |||
| U85772.1 | AAB81564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638165 | Chr.16:21188383 - 21267495 on Build GRCh38 | Gain |
|
| nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain |
|
| esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73539733] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map