Assay Details
Target Gene Details
Entrez Gene ID: | 11060 |
Gene Name: | WW domain containing E3 ubiquitin protein ligase 2 |
Gene Aliases: |
AIP2, WWp2-like |
Location: |
Chr.16:69762284-69941741 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWP2 | NM_001270454.1 | NP_001257383.1 | ||
| NM_001270455.1 | NP_001257384.1 | |||
| NM_007014.4 | NP_008945.2 | |||
| XM_011522823.2 | XP_011521125.1 | |||
| XM_011522825.1 | XP_011521127.1 | |||
| XM_017022879.1 | XP_016878368.1 | |||
| XM_017022881.1 | XP_016878370.1 | |||
| AK312792.1 | ||||
| BC064531.1 | AAH64531.1 | |||
| DC402912.1 | ||||
| JN712744.1 | ||||
| U96114.1 | AAC51325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524364 | Chr.16:69711242 - 69769540 on Build GRCh38 | Loss |
|
| esv3638931 | Chr.16:69738369 - 69799862 on Build GRCh38 | Gain |
|
| esv3892878 | Chr.16:69687512 - 69844258 on Build GRCh38 | Gain |
|
| esv3638930 | Chr.16:69735814 - 69773243 on Build GRCh38 | Gain |
|
| esv3638932 | Chr.16:69762176 - 69924258 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map