Assay Details
Target Gene Details
Entrez Gene ID: | 18 |
Gene Name: | 4-aminobutyrate aminotransferase |
Gene Aliases: |
GABA-AT, GABAT, NPD009 |
Location: |
Chr.16:8674587-8784575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABAT | NM_020686.5 | NP_065737.2 | ||
| AK096146.1 | ||||
| BC015628.2 | AAH15628.1 | |||
| BC031413.1 | AAH31413.1 | |||
| BG749438.1 | ||||
| BI459888.1 | ||||
| BX537362.1 | ||||
| DB443567.1 | ||||
| DB479117.1 | ||||
| L32961.1 | AAA74449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047172 | Chr.16:8528343 - 8810411 on Build GRCh38 | Gain |
|
| nsv1036125 | Chr.16:8670288 - 8704399 on Build GRCh38 | Loss |
|
| esv3570276 | Chr.16:8641724 - 8736165 on Build GRCh38 | Gain |
|
| nsv571409 | Chr.16:8661290 - 8746097 on Build GRCh38 | Gain |
|
| nsv523677 | Chr.16:8670571 - 8685733 on Build GRCh38 | Loss |
|
| nsv571406 | Chr.16:8596737 - 8690609 on Build GRCh38 | Gain |
|
| nsv1037325 | Chr.16:8615311 - 8688379 on Build GRCh38 | Gain |
|
| esv2762154 | Chr.16:8637384 - 8724268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map