Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | NP_001252541.1 | ||
| NM_016284.4 | NP_057368.3 | |||
| NR_049763.1 | ||||
| AB023224.2 | BAA76851.2 | |||
| AF110778.1 | AAF14861.1 | |||
| AK056927.1 | ||||
| AK075125.1 | ||||
| AK092885.1 | ||||
| AK301092.1 | ||||
| AK302495.1 | ||||
| AL117492.1 | CAB55960.1 | |||
| AL832769.1 | ||||
| AL833549.1 | ||||
| BC000779.2 | AAH00779.2 | |||
| BC024317.1 | AAH24317.1 | |||
| BX537840.1 | CAD97851.1 | |||
| CR749237.1 | CAH18093.1 | |||
| EF553522.1 |
More Information
Set Membership: |
|

Genomic Map