Assay Details
Target Gene Details
Entrez Gene ID: | 80063 |
Gene Name: | activating transcription factor 7 interacting protein 2 |
Gene Aliases: |
MCAF2 |
Location: |
Chr.16:10386055-10483640 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7IP2 | NM_001256160.1 | NP_001243089.1 | ||
| NM_024997.3 | NP_079273.2 | |||
| NR_045815.1 | ||||
| NR_045816.1 | ||||
| XM_006720953.3 | XP_006721016.1 | |||
| XM_011522666.2 | XP_011520968.1 | |||
| XM_017023705.1 | XP_016879194.1 | |||
| XM_017023706.1 | XP_016879195.1 | |||
| XM_017023707.1 | XP_016879196.1 | |||
| XM_017023708.1 | XP_016879197.1 | |||
| XM_017023710.1 | XP_016879199.1 | |||
| XM_017023711.1 | XP_016879200.1 | |||
| AK022730.1 | BAB14209.1 | |||
| AK093414.1 | BAC04157.1 | |||
| AK223492.1 | BAD97212.1 | |||
| AK296565.1 | ||||
| AK299040.1 | ||||
| AL832987.1 | CAH56323.1 | |||
| AY560615.1 | AAT66299.1 | |||
| BC033891.1 | AAH33891.1 | |||
| BC069695.1 | AAH69695.1 | |||
| BC069713.1 | AAH69713.1 | |||
| BC069730.1 | AAH69730.1 | |||
| BC137079.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051406 | Chr.16:10365859 - 10570475 on Build GRCh38 | Loss |
|
| nsv571451 | Chr.16:10435543 - 10495488 on Build GRCh38 | Loss |
|
| nsv519263 | Chr.16:10386989 - 10466528 on Build GRCh38 | Gain+Loss |
|
| nsv457405 | Chr.16:10388455 - 10466528 on Build GRCh38 | Loss |
|
| nsv1036128 | Chr.16:10404948 - 10470444 on Build GRCh38 | Loss |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111917498] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map