Assay Details
Target Gene Details
Entrez Gene ID: | 100288072 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 2 |
Gene Aliases: |
- |
Location: |
Chr.16:22162531-22190444 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR42E2 | XM_017023977.1 | XP_016879466.1 | ||
| XM_017023978.1 | 1 | 422 | XP_016879467.1 | |
| XM_017023979.1 | 1 | 422 | XP_016879468.1 | |
| XM_017023980.1 | 1 | 422 | XP_016879469.1 | |
| XM_017023981.1 | 1 | 422 | XP_016879470.1 | |
| XM_017023982.1 | 1 | 422 | XP_016879471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638198 | Chr.16:22121308 - 22167549 on Build GRCh38 | Gain |
|
| nsv469818 | Chr.16:22112125 - 22262126 on Build GRCh38 | Loss |
|
| nsv469628 | Chr.16:22055049 - 22205066 on Build GRCh38 | Gain |
|
| esv3638195 | Chr.16:22082786 - 22273655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map