Assay Details
Target Gene Details
Entrez Gene ID: | 124739 |
Gene Name: | ubiquitin specific peptidase 43 |
Gene Aliases: |
- |
Location: |
Chr.17:9643493-9729687 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP43 | XM_011523641.1 | XP_011521943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833358 | Chr.17:9643743 - 9828246 on Build GRCh38 | Gain |
|
| nsv833357 | Chr.17:9518357 - 9696606 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map