Assay Details
Target Gene Details
Entrez Gene ID: | 23164 |
Gene Name: | myosin phosphatase Rho interacting protein |
Gene Aliases: |
M-RIP, MRIP, RHOIP3, RIP3, p116Rip |
Location: |
Chr.17:17042438-17192648 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPRIP | NM_015134.3 | NP_055949.2 | ||
| NM_201274.3 | NP_958431.2 | |||
| XM_005256563.4 | XP_005256620.1 | |||
| XM_005256564.4 | XP_005256621.3 | |||
| XM_011523761.2 | XP_011522063.1 | |||
| XM_011523762.2 | XP_011522064.1 | |||
| XM_011523763.2 | XP_011522065.1 | |||
| XM_011523764.2 | XP_011522066.1 | |||
| XM_011523765.2 | XP_011522067.1 | |||
| XM_011523766.2 | XP_011522068.2 | |||
| XM_011523767.2 | XP_011522069.2 | |||
| XM_017024393.1 | XP_016879882.1 | |||
| AB098507.1 | BAC78198.1 | |||
| AB189741.1 | BAD89507.1 | |||
| AY296247.1 | AAQ63176.1 | |||
| BC136479.1 | ||||
| BE048871.1 |
More Information
Set Membership: |
|

Genomic Map