Assay Details
Target Gene Details
Entrez Gene ID: | 124872 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyltransferase 2 |
Gene Aliases: |
B4GALT, GALGT2 |
Location: |
Chr.17:49120347-49170722 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALNT2 | NM_001159387.1 | NP_001152859.1 | ||
| NM_001159388.1 | NP_001152860.1 | |||
| NM_153446.2 | NP_703147.2 | |||
| XM_011524314.2 | XP_011522616.1 | |||
| XM_017024173.1 | XP_016879662.1 | |||
| AF510036.1 | AAM34756.1 | |||
| AJ517770.1 | CAD57148.1 | |||
| AJ517771.1 | CAD57149.1 | |||
| AK302876.1 | ||||
| BC113675.1 | ||||
| BC113677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065769 | Chr.17:49091661 - 49191783 on Build GRCh38 | Gain |
|
| nsv1065020 | Chr.17:49130326 - 49149675 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79893555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map