Assay Details
Target Gene Details
Entrez Gene ID: | 256302 |
Gene Name: | N-acetyltransferase domain containing 1 |
Gene Aliases: |
C17orf103, Gtlf3b |
Location: |
Chr.17:21238870-21253266 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NATD1 | NM_152914.2 | 3 | 2493 | NP_690878.2 |
| AK126856.1 | 3 | 2493 | ||
| CR749731.1 | 1 | 663 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain |
|
| nsv953852 | Chr.17:21235688 - 21249887 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map