Assay Details
Target Gene Details
Entrez Gene ID: | 11056 |
Gene Name: | DEAD-box helicase 52 |
Gene Aliases: |
HUSSY19, ROK1 |
Location: |
Chr.17:37609739-37643464 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX52 | NM_001291476.1 | 16 | 6449 | NP_001278405.1 |
| NM_007010.4 | 15 | 6334 | NP_008941.3 | |
| XM_011524232.2 | 15 | 6329 | XP_011522534.1 | |
| XM_011524233.2 | 13 | 6190 | XP_011522535.1 | |
| AK025721.1 | 6 | 2883 |
Target Gene Details
Entrez Gene ID: | 11276 |
Gene Name: | synergin, gamma |
Gene Aliases: |
AP1GBP1, SYNG |
Location: |
Chr.17:37514797-37613637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNRG | XM_017024080.1 | 1 | 3832 | XP_016879569.1 |
| XM_017024081.1 | 1 | 3832 | XP_016879570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833431 | Chr.17:37530641 - 37726517 on Build GRCh38 | Gain |
|
| esv3640494 | Chr.17:37406555 - 37610947 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map