Assay Details
Target Gene Details
Entrez Gene ID: | 55090 |
Gene Name: | mediator complex subunit 9 |
Gene Aliases: |
MED25 |
Location: |
Chr.17:17476986-17493220 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED9 | NM_018019.2 | NP_060489.1 | ||
| AK001055.1 | BAA91483.1 | |||
| BC104988.1 | AAI04989.1 | |||
| BC112029.1 | AAI12030.1 | |||
| HG518168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827909 | Chr.17:17455631 - 17491017 on Build GRCh38 | Gain |
|
| nsv1065710 | Chr.17:17424797 - 17488129 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map