Assay Details
Target Gene Details
Entrez Gene ID: | 22806 |
Gene Name: | IKAROS family zinc finger 3 |
Gene Aliases: |
AIO, AIOLOS, ZNFN1A3 |
Location: |
Chr.17:39757715-39864188 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
More Information
Set Membership: |
|

Genomic Map