Assay Details
Target Gene Details
Entrez Gene ID: | 105371921 |
Gene Name: | uncharacterized LOC105371921 |
Gene Aliases: |
- |
Location: |
Chr.17:80885776-80891713 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371921 | XM_017025502.1 | XP_016880991.1 |
Target Gene Details
Entrez Gene ID: | 57521 |
Gene Name: | regulatory associated protein of MTOR complex 1 |
Gene Aliases: |
KOG1, Mip1 |
Location: |
Chr.17:80544825-80966373 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPTOR | NM_001163034.1 | NP_001156506.1 | ||
| NM_020761.2 | NP_065812.1 | |||
| AB037724.1 | BAA92541.1 | |||
| AB082951.1 | BAC06490.1 | |||
| AK055912.1 | ||||
| AK296321.1 | ||||
| AY090663.1 | AAM09075.1 | |||
| BC136652.1 | ||||
| BC136654.1 | ||||
| GQ183898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952386 | Chr.17:80872501 - 81005700 on Build GRCh38 | Deletion |
|
| esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss |
|
| nsv509676 | Chr.17:80864760 - 80936519 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map