Assay Details
Target Gene Details
Entrez Gene ID: | 147184 |
Gene Name: | transmembrane protein 99 |
Gene Aliases: |
- |
Location: |
Chr.17:40819092-40836274 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM99 | NM_001195386.1 | NP_001182315.1 | ||
| NM_001195387.1 | NP_001182316.1 | |||
| NM_145274.3 | NP_660317.2 | |||
| XM_005257089.4 | XP_005257146.1 | |||
| XM_017024253.1 | XP_016879742.1 | |||
| AK097454.1 | BAC05059.1 | |||
| AK298616.1 | ||||
| BC015365.1 | AAH15365.1 | |||
| BG530583.1 | ||||
| CD689247.1 | ||||
| DB459363.1 | ||||
| DN920898.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79253682] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map