Assay Details
Target Gene Details
Entrez Gene ID: | 9755 |
Gene Name: | TBK1 binding protein 1 |
Gene Aliases: |
ProSAPiP2, SINTBAD |
Location: |
Chr.17:47694065-47712063 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBKBP1 | XM_005257859.4 | XP_005257916.1 | ||
| XM_005257861.4 | XP_005257918.1 | |||
| XM_005257862.3 | XP_005257919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9570 | Chr.17:47693507 - 47702425 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs531909324] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map