Assay Details
Target Gene Details
Entrez Gene ID: | 105371506 |
Gene Name: | uncharacterized LOC105371506 |
Gene Aliases: |
- |
Location: |
Chr.17:5469092-5470360 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371506 | NR_135644.1 | |||
| AW103155.1 |

Genomic Map