Assay Details
Target Gene Details
Entrez Gene ID: | 163 |
Gene Name: | adaptor related protein complex 2 beta 1 subunit |
Gene Aliases: |
ADTB2, AP105B, AP2-BETA, CLAPB1 |
Location: |
Chr.17:35586899-35726417 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2038 | Chr.17:35569617 - 35598796 on Build GRCh38 | Insertion |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map