Assay Details
Target Gene Details
Entrez Gene ID: | 642 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
BH, BMH |
Location: |
Chr.17:30248195-30292166 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLMH | NM_000386.3 | NP_000377.1 | ||
| AF091082.1 | AAC72951.1 | |||
| AK223403.1 | BAD97123.1 | |||
| AK301948.1 | ||||
| AK312896.1 | ||||
| BC003616.2 | AAH03616.1 | |||
| BT007018.1 | AAP35664.1 | |||
| X92106.1 | CAA63078.1 |
More Information
Set Membership: |
|

Genomic Map