Assay Details
Target Gene Details
Entrez Gene ID: | 101928855 |
Gene Name: | uncharacterized LOC101928855 |
Gene Aliases: |
- |
Location: |
Chr.17:80801640-80805632 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928855 | NR_110852.1 | 4 | 2006 | |
| AK123620.1 | 4 | 2006 |
Target Gene Details
Entrez Gene ID: | 57521 |
Gene Name: | regulatory associated protein of MTOR complex 1 |
Gene Aliases: |
KOG1, Mip1 |
Location: |
Chr.17:80544825-80966373 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPTOR | NM_001163034.1 | NP_001156506.1 | ||
| NM_020761.2 | NP_065812.1 | |||
| AB037724.1 | BAA92541.1 | |||
| AB082951.1 | BAC06490.1 | |||
| AK055912.1 | ||||
| AK295964.1 | ||||
| AK296321.1 | ||||
| AY090663.1 | AAM09075.1 | |||
| BC064515.1 | AAH64515.1 | |||
| BC136652.1 | ||||
| BC136654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss |
|
| nsv833563 | Chr.17:80605936 - 80813742 on Build GRCh38 | Gain |
|
| nsv576164 | Chr.17:80768726 - 80871552 on Build GRCh38 | Gain |
|
| nsv509675 | Chr.17:80740098 - 80806572 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map